Learn More
Med Vet International Heavy Duty Chrome Hanger for X-Ray Apron, J0676H, Each
Catalog No. 5000495890
Encompass_Preferred
Heavy duty chrome hanger: chrome plate 3/16” recommended to protect x-ray aprons and increase product life. Hanger will hold up to 50lb (22.7kg)
Catalog No. 50-004-95890 Supplier Med Vet International Supplier No. JOR676H
Heavy duty chrome hanger: chrome plate 3/16” recommended to protect x-ray aprons and increase product life. Hanger will hold up to 50lb (22.7kg)