Learn More
Alkali Scientific ASI™ Hanging Cell Culture Inserts with TC Treated Plates, Polycarbonate (PC), 6 Inserts / 6well Plate, PS,PC membrance, Translucent, 24mmØ ,3.00μm pore, 1.50-2.50mL, 4.52cm², Sterile to SAL 10-6, 6/plate/24/case
SDP
Catalog No. 503028617
Encompass_Preferred
ASI™ Hanging Cell Culture Inserts ▪ Distance from membrane to the bottom of the well: 6well Insert 1.2 mm,12well Insert 1.2 mm, 24well Insert 1.3 mm▪ Nominal membrane thickness: 10 ~ 23 μm▪ Frame material: Polystyrene (PS)▪ Packed in cell culture-treated cell culture plates ▪ Also known as Transwell permeable support inserts▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-8617 Supplier Alkali Scientific Supplier No. TCH0630
ASI™ Hanging Cell Culture Inserts ▪ Distance from membrane to the bottom of the well: 6well Insert 1.2 mm,12well Insert 1.2 mm, 24well Insert 1.3 mm▪ Nominal membrane thickness: 10 ~ 23 μm▪ Frame material: Polystyrene (PS)▪ Packed in cell culture-treated cell culture plates ▪ Also known as Transwell permeable support inserts▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free