Learn More
Grainger Hanging Glass Rack 16 in L Gold
Catalog No. 502763660
Encompass_Preferred
The Grainger 16W524 is a Philips 16-watt T8 fluorescent light bulb that provides 1600 lumens of light output. This 4 foot long bulb has a medium bi-pin base and a color temperature of 4100K. It has an average rated life of 20,000 hours and meets EISA standards for energy efficiency. The Philips 16W524 T8 is designed for use in commercial and residential fluorescent fixtures and provides bright, white light.
Catalog No. 50-276-3660 Supplier Grainger Supplier No. 16W524
The Grainger 16W524 is a Philips 16-watt T8 fluorescent light bulb that provides 1600 lumens of light output. This 4 foot long bulb has a medium bi-pin base and a color temperature of 4100K. It has an average rated life of 20,000 hours and meets EISA standards for energy efficiency. The Philips 16W524 T8 is designed for use in commercial and residential fluorescent fixtures and provides bright, white light.