Learn More
Grainger Hanging Glass Rack 24 in L Silver
Catalog No. 502764841
Encompass_Preferred
The Crestware Hanging Glass Rack is a 24 inch wide stainless steel wire rack designed for drying stemware, glassware, and dishware. It features two tiers of closely spaced wires to hold glasses securely upside down for drip drying. The rack can hold up to 18 glasses. It includes 4 mounting hooks to hang from under a cabinet or shelf. This commercial grade rack is made from heavy gauge 201 stainless steel for durability and rust resistance. Its open wire design allows for maximum air circulation and drying. Overall dimensions are 24"W x 20"H x 14"D.
Catalog No. 50-276-4841 Supplier Grainger Supplier No. 16W519
The Crestware Hanging Glass Rack is a 24 inch wide stainless steel wire rack designed for drying stemware, glassware, and dishware. It features two tiers of closely spaced wires to hold glasses securely upside down for drip drying. The rack can hold up to 18 glasses. It includes 4 mounting hooks to hang from under a cabinet or shelf. This commercial grade rack is made from heavy gauge 201 stainless steel for durability and rust resistance. Its open wire design allows for maximum air circulation and drying. Overall dimensions are 24"W x 20"H x 14"D.