Learn More
IBI SCIENTIFIC 3P Hanging Pipette Stand, 4 Place w/ Filler, Right Side
Catalog No. NC0605532
Encompass_Preferred
Hanging Pipette Stand. IBI Scientific. 4-place pipette holder with an additional place for a mechanical pipette filler (on right side.) Constructed of 1/4-inch cast acrylic. Easily attaches to wall or fume hood with double-sided tape.
Catalog No. NC0605532 Supplier IBI SCIENTIFIC 3P Supplier No. PST50
Hanging Pipette Stand. IBI Scientific. 4-place pipette holder with an additional place for a mechanical pipette filler (on right side.) Constructed of 1/4-inch cast acrylic. Easily attaches to wall or fume hood with double-sided tape.