Learn More
SCALESPLUS HANGING SCALE 30 LB X 0.02 LB
SDP
Catalog No. NC2385864
Encompass
The Class III NTEP AHS-30 hanging scale from Intelligent Weighing is the perfect solution for weighing. This digital hanging scale is battery operated and light weight for portable weighing applications. An optional hanging pan extends the AHS to other weighing applications. The AHS-30 is fully compliant with NTEP regulations and is legal for trade. The Certificate of Conformance number is 18-098.Lightweight and simple to operateBattery powered-4 size “C” type batteries (not included)Battery power allows for up to 250 hours of operationPower Adapter option (not included) - If adapter is used then remove batteries1500 divisionsConstructed of ABS plasticSafe overload up 200% capacityWeight reading stabilizes in less than 2 secondsAuto zero tracking1 year warrantyDoes not weigh in grams, only weighs in lb and kg
Catalog No. NC2385864 Supplier SCALESPLUS Supplier No. AHS30
The Class III NTEP AHS-30 hanging scale from Intelligent Weighing is the perfect solution for weighing. This digital hanging scale is battery operated and light weight for portable weighing applications. An optional hanging pan extends the AHS to other weighing applications. The AHS-30 is fully compliant with NTEP regulations and is legal for trade. The Certificate of Conformance number is 18-098.Lightweight and simple to operateBattery powered-4 size “C” type batteries (not included)Battery power allows for up to 250 hours of operationPower Adapter option (not included) - If adapter is used then remove batteries1500 divisionsConstructed of ABS plasticSafe overload up 200% capacityWeight reading stabilizes in less than 2 secondsAuto zero tracking1 year warrantyDoes not weigh in grams, only weighs in lb and kg