Learn More
Electron Microscopy Sciences Hank's Balanced Salts Formula I 1 LT
SDP
Catalog No. 50980254
Encompass_Preferred
All of our Hanks' Balanced Salt Solutions contain: Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All solutions are prepared in 1X strength.
Catalog No. 50-980-254 Supplier Electron Microscopy Sciences Supplier No. 1176810
All of our Hanks' Balanced Salt Solutions contain: Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All solutions are prepared in 1X strength.