Learn More
Electron Microscopy Sciences Hank's Balanced Salts Formula III 500 ML
SDP
Catalog No. 50980257
Encompass_Preferred
All of our Hanks' Balanced Salt Solutions contain: Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All solutions are prepared in 1X strength.
Catalog No. 50-980-257 Supplier Electron Microscopy Sciences Supplier No. 1177005
All of our Hanks' Balanced Salt Solutions contain: Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application. All solutions are prepared in 1X strength.