Learn More
Supelco Inc HANKS BALANCE SALT 500ML
Catalog No. NC0319985
Encompass
HanksAE Balanced Salt, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Catalog No. NC0319985 Supplier Supelco Inc Supplier No. H6648500ML
HanksAE Balanced Salt, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture