Learn More
STEMCELL Technologies HBSS with 10 mM HEPES, Without Phenol Red, Size: 500 mL
SDP
Catalog No. NC9162583
Encompass
Hanks’ Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating, or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions, and a carbohydrate as an energy source. Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.
Catalog No. NC9162583 Supplier STEMCELL Technologies Supplier No. 37150
Hanks’ Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating, or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions, and a carbohydrate as an energy source. Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.