Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanks' Balanced Salt solution Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture | 100ML
Catalog No. NC2117850
Encompass
Hanks' Balanced Salt solution Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture | 100ML
Catalog No. NC2117850 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H6648100ML
Hanks' Balanced Salt solution Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture | 100ML