Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanks Balanced Salt solut500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H82646X500ML
Encompass
Hanks Balanced Salt SOLUTIONon (HBSS) is a buffer that mimics biological medium and stabilizes cells. It is useful in leucocyte labeling studies. HBSS is an effective preservative for periodontal ligament cells. HBSS lacks D-glucose and essential inorganic ions and is useful in starvation studies.
Catalog No. 50-290-1441