Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanks Balanced Salt soluti1L
Supplier: Sigma Aldrich Fine Chemicals Biosciences H82641L
Hanks Balanced Salt SOLUTIONon (HBSS) is a buffer that mimics biological medium and stabilizes cells. It is useful in leucocyte labeling studies. HBSS is an effective preservative for periodontal ligament cells. HBSS lacks D-glucose and essential inorganic ions and is useful in starvation studies.