Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanks' Balanced Salts, Without Sodium Bicarbonate, MP Biomedicals™

Catalog No. MP091710122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091710122 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091710122 Supplier MP Biomedicals, Inc Supplier No. 091710122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hanks' Balanced Salts, Without Sodium Bicarbonate, Orange Powder

  • Hanks' Balanced Salts solution is used to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
pH 6.0 to 7.0 (before adjustment with sodium bicarbonate)
Physical Form Powder
Quantity 1 x 10 L
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.