Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanks’ Balanced Salt Solution 10x, 500 mL
Catalog No. NC1140338
Encompass
Calcium and magnesium support cell adhesion. Recommended for suspension cultures or whenever cell clumping is a problem.
Catalog No. NC1140338 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H4641500ML
Calcium and magnesium support cell adhesion. Recommended for suspension cultures or whenever cell clumping is a problem.