Learn More
Andwin Scientific Checker Plus pH Tester + Gloves
SDP
Catalog No. NC1360217
Encompass_Preferred
The HI98100 Checker Plus is the NEXT GENERATION of the original Hanna Checker pH tester. The Checker is the most popular pH meter in the world with over 1 million meters used since its introduction in 1991. From entry-level growers to professional horticulturalists, the Checker has been helping people with their pH measurement as a meter that is simple to use and operate. The Checker Plus now offers automatic calibration to one or two points, automatic buffer recognition, calibrated buffer tags, stability indicator, low battery indicator, and selectable automatic shut off. Plastic Carrying Case - The HI98100 Checker Plus is supplied complete with meter, probe, calibration solutions and cleaning solutions packaged in a durable plastic carrying case.
Catalog No. NC1360217 Supplier Andwin Scientific Supplier No. HI98100
The HI98100 Checker Plus is the NEXT GENERATION of the original Hanna Checker pH tester. The Checker is the most popular pH meter in the world with over 1 million meters used since its introduction in 1991. From entry-level growers to professional horticulturalists, the Checker has been helping people with their pH measurement as a meter that is simple to use and operate. The Checker Plus now offers automatic calibration to one or two points, automatic buffer recognition, calibrated buffer tags, stability indicator, low battery indicator, and selectable automatic shut off. Plastic Carrying Case - The HI98100 Checker Plus is supplied complete with meter, probe, calibration solutions and cleaning solutions packaged in a durable plastic carrying case.