Learn More
Andwin Scientific Digital Glass Body pH Electrode for General Purpose
SDP
Catalog No. NC1247851
Encompass
The HI11310 is a combination, glass, refillable pH electrode with a built-in temperature sensor. This probe features a 3.5 mm connector designed exclusively to be used with Hanna Instruments edge Tablet Meters.
Catalog No. NC1247851 Supplier Andwin Scientific Supplier No. HI11310
The HI11310 is a combination, glass, refillable pH electrode with a built-in temperature sensor. This probe features a 3.5 mm connector designed exclusively to be used with Hanna Instruments edge Tablet Meters.