Learn More
HANNA INSTRUMENTS INC Industrial Flat Tip pH Electrodes
SDP
Catalog No. NC1824129
Encompass
Hanna presents a series of combination pH electrodes incorporating over 20 years of electrode manufacturing experience. The most advanced feature of this series is the electrode shape with a flat tip, virtually eliminating deposits that can foul the electrode, significantly reducing necessary maintenance. This characteristic makes flat tip electrodes ideal for continuous in-line monitoring and for solutions containing aggressive chemicals.Self-cleaning flat tip sensor for significantly reduced maintenance.Provided with an internal matching pin that can avoid typical problems caused by grounding loop current.3/4” NPT external thread on both ends for easy installation.
Catalog No. NC1824129 Supplier HANNA INSTRUMENTS INC Supplier No. HI10062005
Hanna presents a series of combination pH electrodes incorporating over 20 years of electrode manufacturing experience. The most advanced feature of this series is the electrode shape with a flat tip, virtually eliminating deposits that can foul the electrode, significantly reducing necessary maintenance. This characteristic makes flat tip electrodes ideal for continuous in-line monitoring and for solutions containing aggressive chemicals.Self-cleaning flat tip sensor for significantly reduced maintenance.Provided with an internal matching pin that can avoid typical problems caused by grounding loop current.3/4” NPT external thread on both ends for easy installation.