Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hansa Yellow 45.0+%, TCI America™
SDP

Catalog No. H031225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H031225G Supplier TCI America Supplier No. H031225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2512-29-0
Molecular Formula C17H16N4O4
MDL Number MFCD00078234
Synonym Pigment Yellow 1

Specifications

Quantity 25 g
Formula Weight 340.34
Percent Purity ≥45.0% (E)
Physical Form Crystalline Powder
Chemical Name or Material Hansa Yellow
EINECSNumber (5)-3149
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.