Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanus solution, Solstice

Catalog No. 6000631 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-31 500 mL
60-006-30 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-006-31 Supplier Solstice Supplier No. 35097500ML

Volumetric, 0.1 M IBr

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Quantity 500 mL
UN Number UN2789
Beilstein 3587194
Flash Point 40°C (104°F)
Formula Weight 206.81g/mol
Chemical Name or Material Hanus solution
Hazard Statement P280-P305 + P351 + P338-P310
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.