Learn More
Aviva Systems Biology Corp HAO2 Antibody - N-terminal region (ARP41434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HAO2. Alternate names: GIG16, HAOX2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57852