Learn More
Promocell GmbH PromoCell HAoAF-c 500,000 Human Aortic Adventitial Fibroblasts, cryopreserved
Supplier:  Promocell GmbH C12380
Encompass
Primary Human Aortic Adventitial Fibroblasts (HAoAF) are isolated from human aortic adventitial tissue. HAoAF can be used for studies concerning vascular diseases e.g. neointimal formation, vascular remodeling, atherosclerosis, and hypertension. They are also often used for the examination of fibroblast disorders like fibrosis or other diseases linked to either imperfect or excessive accumulation of fibroblasts.
Catalog No. 50-306-010