Learn More
Promocell GmbH PromoCell HAoEC-c 500,000 Human Aortic Endothelial Cells, cryopreserved
Catalog No. 50305990
Encompass
Primary Human Aortic Endothelial Cells (HAoEC) are isolated from the human ascending (thoracic) and descending (abdominal) aorta. They are useful for studying vascular diseases such as thrombosis, atherosclerosis, and hypertension as well as for stent-graft compatibility testing.
Catalog No. 50-305-990 Supplier Promocell GmbH Supplier No. C12271
Primary Human Aortic Endothelial Cells (HAoEC) are isolated from the human ascending (thoracic) and descending (abdominal) aorta. They are useful for studying vascular diseases such as thrombosis, atherosclerosis, and hypertension as well as for stent-graft compatibility testing.