Learn More
Aviva Systems Biology Corp HAPLN2 Antibody - middle region (ARP57560_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57560_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAPLN2. It was validated on Western Blot. HAPLN2,hyaluronan and proteoglycan link protein 2,BRAL1; HAPLN2,hyaluronan and proteoglycan link protein 2,hyaluronan and proteoglycan link protein 2,OTTHUMP00000031988,brain link protein 1,brain link protein-1The immunogen is a synthetic peptide directed towards the middle region of human HAPLN2. Alternate names: BRAL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-286-332