Learn More
Aviva Systems Biology Corp HAPLN4 Antibody - C-terminal region (ARP49777_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAPLN4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HAPLN4. Alternate names: BRAL2, KIAA1926ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-448