Learn More
Boca Scientific Inc Happy Cell ASM formulated in MEM, 4X Concentrate, 25ml
Catalog No. 501685628
Encompass
Happy Cell ASM is a unique, low viscosity, low density, cell culture reagent that permanently suspends cells. It allows easy labeling, washing and dosing of cells. This product was developed for drug discovery and biomedical research. It has proven to support 3D growth in cancer, stem and primary cells, and allows for rapid tumor spheroid formation. Happy Cell ASM is suitable for high content imaging and automated liquid handling. This version of Happy Cell ASM is formulated in MEM as a 4X concentrate.
Catalog No. 50-168-5628 Supplier Boca Scientific Inc Supplier No. VHCME25
Happy Cell ASM is a unique, low viscosity, low density, cell culture reagent that permanently suspends cells. It allows easy labeling, washing and dosing of cells. This product was developed for drug discovery and biomedical research. It has proven to support 3D growth in cancer, stem and primary cells, and allows for rapid tumor spheroid formation. Happy Cell ASM is suitable for high content imaging and automated liquid handling. This version of Happy Cell ASM is formulated in MEM as a 4X concentrate.