Learn More
Boca Scientific Inc Happy Cell ASM General Purpose & Scale Up Kit - RPMI: 1 x 25ml Happy Cell ASM RPMI 4X Concentrate, 1 x vial Inactivation Solution, 5 x low attachment 96 well microplates, 2 x 50ml bioreactor tubes
Catalog No. 501685632
Encompass
The complete Happy Cell ASM General Purpose & Scale-Up Kit provides the essential tools for growing 3D cells and micro-tissues to a larger scale. This kit includes Happy Cell ASM formulated in RPMI (4x concentrate, 25ml), Happy Cell Inactivation Solution, untreated low attachment 96 well microplates, and 50ml bioreactor tubes with vented caps. The included Happy Cell concentrate is sufficient for up to 1000 experiments in a 96 well microplate format or up to 4000 experiments in a 384 well microplate format. Happy Cell ASM is a unique, low viscosity, low density, cell culture reagent that permanently suspends cells. It allows easy labeling, washing and dosing of cells, and is suitable for high content imaging and automated liquid handling. Happy Cell ASM Inactivation Solution is a liquid reagent designed to enhance the performance of Happy Cell ASM and improve the quality of your data.
Catalog No. 50-168-5632 Supplier Boca Scientific Inc Supplier No. VHCK2 RPMI
The complete Happy Cell ASM General Purpose & Scale-Up Kit provides the essential tools for growing 3D cells and micro-tissues to a larger scale. This kit includes Happy Cell ASM formulated in RPMI (4x concentrate, 25ml), Happy Cell Inactivation Solution, untreated low attachment 96 well microplates, and 50ml bioreactor tubes with vented caps. The included Happy Cell concentrate is sufficient for up to 1000 experiments in a 96 well microplate format or up to 4000 experiments in a 384 well microplate format. Happy Cell ASM is a unique, low viscosity, low density, cell culture reagent that permanently suspends cells. It allows easy labeling, washing and dosing of cells, and is suitable for high content imaging and automated liquid handling. Happy Cell ASM Inactivation Solution is a liquid reagent designed to enhance the performance of Happy Cell ASM and improve the quality of your data.