Learn More
Boca Scientific Inc Happy Cell ASM Starter Kit DMEM: Happy Cell ASM DMEM, Inactivation Solution, Low Attachment 96 Well Plate
Supplier:  Boca Scientific Inc VHCK1DMEM
Encompass
This starter kit includes Happy Cell ASM formulated in DMEM (4x concentrate, 2.5ml), Happy Cell Inactivation Solution, and a low attachment 96 well microplate. This product is intended as a trial kit to understand the properties of Happy Cell and how to optimize for its use. The included Happy Cell ASM is sufficient for up to 100 experiments at 100 microliters per experiment. Happy Cell ASM is a unique, low viscosity, low density, cell culture reagent that permanently suspends cells. It allows easy labeling, washing and dosing of cells, and is suitable for high content imaging and automated liquid handling. Happy Cell ASM Inactivation Solution is a liquid reagent designed to enhance the performance of Happy Cell ASM and improve the quality of your data. It facilitates the recovery of 3D multi-cellular structures by irreversibly disrupting the Happy Cell ASM suspension polymer complex without compromising cellular integrity and function.
Catalog No. 50-168-5633