Learn More
Carbosynth LLC. Haptoglobin Heavy Tryptic Peptide Standard, 4nmol
SDP
Catalog No. NC2947974
Encompass_Preferred
This Haptoglobin Heavy Tryptic Peptide Standard has the potential for use in proteomics studies and in the identification and quantitation of proteins. Haptoglobin is involved in the removal of free hemoglobin and also in neutralizing oxidative damage.
Catalog No. NC2947974 Supplier Carbosynth LLC. Supplier No. BP178751PIECE
This Haptoglobin Heavy Tryptic Peptide Standard has the potential for use in proteomics studies and in the identification and quantitation of proteins. Haptoglobin is involved in the removal of free hemoglobin and also in neutralizing oxidative damage.