Learn More
Alpco Ltd Equine Haptoglobin ELISA
SDP
Supplier:  Alpco Ltd 41HAPEQE01
Encompass
The Equine Haptoglobin ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Haptoglobin in horse biological samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3577