Learn More
Athens Research And Technology Inc HAPTOGLOBIN HU 1MG
Supplier:  Athens Research And Technology Inc 16160801161MG
Encompass_Preferred
Haptoglobin, Human Plasma: 1 MG; Lyophilized salt-free solid; MW Hp 1-1: 86,000; Hp 2-1: 200,000; Hp 2-2: 400,000; EC=1.20; Purity:> =95% by SDS-PAGE. Protein is prepared & purified from human plasma in the Athens Research & Technology laboratory.
Catalog No. 50-489-744