Learn More
Aviva Systems Biology Corp Harbi1 Antibody - N-terminal region (ARP55707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Harbi1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: D230010M03Rik, RP23-12H6.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-068