Learn More
TA Instruments Hard Anodized Aluminum, 20 mm, Quick Change Plate, HR X0, DHR, ARES-G2
Catalog No. 5000580873 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2 Hard Anodized Aluminum Quick Change Plate (20 mm) provides a durable, lightweight option for use with Advanced Peltier Plates. Designed for secure placement, consistent sample contact, and precise rheological measurements.
Catalog No. 50-005-80873 Supplier TA Instruments Supplier No. 402493.901
Waters TA Instruments HR X0, DHR, ARES-G2 Hard Anodized Aluminum Quick Change Plate (20 mm) provides a durable, lightweight option for use with Advanced Peltier Plates. Designed for secure placement, consistent sample contact, and precise rheological measurements.