Learn More
TA Instruments Hard Anodized Aluminum, 25 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580822 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Quick Change Plate (25 mm) is compatible with Advanced Peltier Plates and ARES-G2 APS systems. Features lightweight, durable construction and precise sample alignment for reproducible measurements.
Catalog No. 50-005-80822 Supplier TA Instruments Supplier No. 402630.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Quick Change Plate (25 mm) is compatible with Advanced Peltier Plates and ARES-G2 APS systems. Features lightweight, durable construction and precise sample alignment for reproducible measurements.