Learn More
TA Instruments Hard Anodized Aluminum, 40 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Supplier:  TA Instruments 402631.901
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Quick Change Plate (40 mm) is designed for use with Advanced Peltier Plates and ARES-G2 APS Standard Flat Plate. Lightweight, durable, and ensures consistent sample placement and measurement accuracy.
Catalog No. 50-005-80751