Learn More
TA Instruments Hard Anodized Aluminum, 42 mm Dia., 15 mm Edge Height, Immersion Cup, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580754 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Immersion Cup (42 mm diameter, 15 mm edge height) includes two removable sidewalls (one aluminum and one clear polycarbonate) for easy sample loading and trimming.
Catalog No. 50-005-80754 Supplier TA Instruments Supplier No. 402681.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Immersion Cup (42 mm diameter, 15 mm edge height) includes two removable sidewalls (one aluminum and one clear polycarbonate) for easy sample loading and trimming.