Learn More
TA Instruments Hard Anodized Aluminum, 50 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580802 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Quick Change Plate (50 mm) provides reliable placement for Advanced Peltier Plates and ARES-G2 APS systems. Lightweight, durable, and ensures reproducible rheological testing.
Catalog No. 50-005-80802 Supplier TA Instruments Supplier No. 402632.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Hard Anodized Aluminum Quick Change Plate (50 mm) provides reliable placement for Advanced Peltier Plates and ARES-G2 APS systems. Lightweight, durable, and ensures reproducible rheological testing.