Learn More
Med Vet International Hard-back Orthopedic Saw, 140mm long x 23mm deep with 0.4mm cut, J0219X, Each
Catalog No. 5000585388
Encompass_Preferred
Stainless steel saw with ridged back and is autoclavable. The stiff back helps guide the saw compared to hacksaw-type blades, which often deviate. 140mm long x 23mm deep with 0.4mm cut.
Catalog No. 50-005-85388 Supplier Med Vet International Supplier No. JOR219X
Stainless steel saw with ridged back and is autoclavable. The stiff back helps guide the saw compared to hacksaw-type blades, which often deviate. 140mm long x 23mm deep with 0.4mm cut.