Learn More
3M™ Hard Hat Mounted Task Light, 150 Lumen
Catalog No. 19153701 Shop All 3M Products
Change view
Click to view available options
Quantity:
10/Cs.
Catalog No. Quantity
19-153-701 10/Cs.
1 options
Catalog No. 19-153-701 Supplier 3M™ Supplier No. 7010316628
Add to Cart
Add to Cart
Hard Hat Mounted Task Light is a water resistant (IPX-4 rated) hard hat mounted head lamp is ideal for industrial lighting applications
  • Lightweight
  • IPX-4 Water resistant
  • 4 modes
  • Comes with 3M™ Dual Lock fasteners eliminating the need for a headband that could interfere with earmuffs and other attachments
  • Comes with a rubber headband which provides a secure attachment to most 3M hard hats
  • Comes with nylon headband which allows the lamp to be worn by the user without wearing a hard hat
  • Versatile headlamp that allows hands free lighting for any industrial worker
TRUSTED_SUSTAINABILITY
Specifications
Battery Life Runs up to 150 hours on low flood setting and 28 hours on high spot setting
Color Brown
Includes 3 AAA Batteries, 3M™ Dual Lock fasteners , Rubber headband, Nylon headband
Model HL200
Product Type 150 Lumen Spot Light
Quantity 10/Cs.
Weight (Metric) 83 g