Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Hard Hat Shade

Catalog No. 19555123 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Blue
Orange
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-123 White
19-555-125 Orange
19-555-126 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-555-123 Supplier Pyramex Safety Products Supplier No. HPSHADE10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pyramex Safety™ Hard Hat Shade blocks sun from the face and neck on job sites lacking other forms of shade

  • Soft lightweight material
  • Hi-vis material for greater visibility
  • Fits on the outside of cap style and full brim hard hats

Specifications

Color White
For Use With (Equipment) Hard Hat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.