Learn More
Bio Rad Laboratories HARD-SHELL FULL-HEIGHT 96-WELL
Catalog No. 509011584
Encompass
PCR plates, 96-well; Hard-Shell; Bio-Rad; Semi-skirted; Full-height; Clear shell/clear well; Polypropylene; Raised rims; Uniform wells; Thin-wall; Black lettering; DNase-, RNase-, and human DNA-free; 25
Catalog No. 50-901-1584 Supplier Bio Rad Laboratories Supplier No. HSS9601
PCR plates, 96-well; Hard-Shell; Bio-Rad; Semi-skirted; Full-height; Clear shell/clear well; Polypropylene; Raised rims; Uniform wells; Thin-wall; Black lettering; DNase-, RNase-, and human DNA-free; 25