Learn More
Bio Rad Laboratories HARD-SHELL THIN-WALL 384-WELL
Catalog No. 509011600
Encompass
PCR plates, 384-well Hard-Shell Bio-Rad Skirted Clear shell White well Polypropylene Raised rims Uniform wells Thin-wall Free of DNase, RNase, and human DNA 50
Catalog No. 50-901-1600 Supplier Bio Rad Laboratories Supplier No. HSP3805
PCR plates, 384-well Hard-Shell Bio-Rad Skirted Clear shell White well Polypropylene Raised rims Uniform wells Thin-wall Free of DNase, RNase, and human DNA 50