Learn More
Bio Rad Laboratories HARD-SHELL THIN-WALL 96-WELL S
Supplier:  Bio Rad Laboratories HSP9601B
Encompass
Plate, PCR; Bio-Rad; 96-well; Skirted; Hard-Shell; Low-profile; White shell; Clear well; PP; Raised rims; Uniform wells; Thin-wall; Black lettering; Free of DNase, RNase, and human DNA; Recommended vol.: 5 to 125uL; bulk; 400 plates/Pk
Catalog No. 50-901-1846