Learn More
Bio Rad Laboratories HARD-SHELL THIN-WALL 96-WELL S
Supplier:  Bio Rad Laboratories HSP9601
Encompass
PCR Plate, 96-Well Skirted Bio-Rad Hard-Shell Low profile Thin wall for fast PCR Clear well Optimized for low-volume reactions Skirt, deck molded from rigid thermostable polymer Free of DNase, RNase, human DNA White shell 50
Catalog No. 50-901-0022