Learn More
Bio Rad Laboratories HARD-SHELL THIN-WALL 96-WELL S
Supplier:  Bio Rad Laboratories HSP9901
Encompass
PCR plates, 96-well Hard-Shell Bio-Rad Skirted Low-profile White shell Clear well 7-digit sequential Code 39 PP Raised rims Uniform wells Thin-wall Black lettering Free of Dnase/RNase, human DNA Recommended vol. 5-125uL
Catalog No. 50-901-1807