Learn More
Bio Rad Laboratories HARD-SHELL THIN-WALL 96-WELL S
Supplier:  Bio Rad Laboratories HSP9611
Encompass
PCR plates, 96-well Hard-Shell Bio-Rad Skirted Low-profile Red shell Clear well Polypropylene Raised rims Uniform wells Thin-wall Black lettering Free of DNase, RNase, and human DNA Recommended vol. 5 to 125uL 50
Catalog No. 50-901-1622