Learn More
Medicus Health Hardside Specimen Transport Cooler, Medium, 19 Quart
SDP
Catalog No. NC1070907
Encompass
∙ 19 Quart Specimen Transport Cooler, Medium are perfect for transporting specimens to the lab and maintaining temperatures ∙Includes attached prominent sticker Biohazard & Specimen Only stickers∙Great option for Labs or Blood Banks to send units using specimens to the ER or Surgery to help prevent units from returning over temperature∙Stainless Steel hardware will provide many years of durable service∙Convenient handle on top and heavy duty strap on this medical transport cooler can be used to secure lid during transport or be used as a shoulder strap∙Constructed of durable polypropylene for easy cleaning∙Insulated with High-Grade molded Polystyrene Foam∙Non-absorbent surface that's easy to clean ∙Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙Internal Dimensions: 13.5"L x 8"W x 10"H; External Dimensions: 16.6"L x 11"W x 13"H
Catalog No. NC1070907 Supplier Medicus Health Supplier No. 5319M1
∙ 19 Quart Specimen Transport Cooler, Medium are perfect for transporting specimens to the lab and maintaining temperatures ∙Includes attached prominent sticker Biohazard & Specimen Only stickers∙Great option for Labs or Blood Banks to send units using specimens to the ER or Surgery to help prevent units from returning over temperature∙Stainless Steel hardware will provide many years of durable service∙Convenient handle on top and heavy duty strap on this medical transport cooler can be used to secure lid during transport or be used as a shoulder strap∙Constructed of durable polypropylene for easy cleaning∙Insulated with High-Grade molded Polystyrene Foam∙Non-absorbent surface that's easy to clean ∙Airtight closure seal is provided by an EVA Gasket Seal recessed into the lid∙Internal Dimensions: 13.5"L x 8"W x 10"H; External Dimensions: 16.6"L x 11"W x 13"H