Learn More
Medicus Health Hard Side Ice Pack, 16 oz.
SDP
Catalog No. NC2608680
Encompass
∙ Hard Side Ice Packs, 16 oz, sold by the each ∙ Great for specimen transport applications ∙ Dimensions: 7"L x 4" W x 1.5"D∙ Rigid, puncture-resistant durable hdpe ice packs provide excellent protection in cold chain applications ∙ Reusable freezer packs are used to transport temperature sensitive items safely and more efficiently than ice∙ Used in coolers and ice chests∙ Freeze before use∙ Melting/freezing point of 0 degrees C∙ Reusable. non-toxic gel that lasts for years
Catalog No. NC2608680 Supplier Medicus Health Supplier No. 5318M7
∙ Hard Side Ice Packs, 16 oz, sold by the each ∙ Great for specimen transport applications ∙ Dimensions: 7"L x 4" W x 1.5"D∙ Rigid, puncture-resistant durable hdpe ice packs provide excellent protection in cold chain applications ∙ Reusable freezer packs are used to transport temperature sensitive items safely and more efficiently than ice∙ Used in coolers and ice chests∙ Freeze before use∙ Melting/freezing point of 0 degrees C∙ Reusable. non-toxic gel that lasts for years