Learn More
MSC Sanitaire Hard Floor Cleaner for HydroClean Washer, Concentrated, 1 gal
Catalog No. 502815914
Encompass
This Sanitaire Hard Floor Cleaner is a concentrated, clear water-white liquid with a characteristic fragrance, designed for use with the HydroClean® Hard Floor Washer (SC930). It is a solvent-based formula with a pH range of 7.0-9.0.
  • Formulated for use with compatible hard floor washers
  • One gallon (128 oz) bottle
  • Concentrated formula, mixes one part solution with seven parts warm tap water
  • Specific mixing ratio for tank: 5 oz solution to 30 oz water
  • Solvent-based composition
  • Ideal for sealed hard floor applications
  • Maintains a pH range of 7.0-9.0
  • Contains C9-C11 Alcohol Ethoxylate and Linear Alcohol Ethoxylate (<1%)
  • Contains 0% VOC content
Catalog No. 50-281-5914 Supplier MSC Supplier No. 20646444
This Sanitaire Hard Floor Cleaner is a concentrated, clear water-white liquid with a characteristic fragrance, designed for use with the HydroClean® Hard Floor Washer (SC930). It is a solvent-based formula with a pH range of 7.0-9.0.
  • Formulated for use with compatible hard floor washers
  • One gallon (128 oz) bottle
  • Concentrated formula, mixes one part solution with seven parts warm tap water
  • Specific mixing ratio for tank: 5 oz solution to 30 oz water
  • Solvent-based composition
  • Ideal for sealed hard floor applications
  • Maintains a pH range of 7.0-9.0
  • Contains C9-C11 Alcohol Ethoxylate and Linear Alcohol Ethoxylate (<1%)
  • Contains 0% VOC content