Learn More
Revvity Health Sciences Inc Hard Tissue Homogenizing Mix 2.8 mm Ceramic (2 mL Reinforced Tubes) - Microbial DNA Free (50/pk)
Catalog No. 502809099
Encompass
Hard Tissue Homogenizing Mix 2.8 mm Ceramic (2 mL Reinforced Tubes) - Microbial DNA Free (50/pk)
Catalog No. 50-280-9099 Supplier Revvity Health Sciences Inc Supplier No. 19628D
Hard Tissue Homogenizing Mix 2.8 mm Ceramic (2 mL Reinforced Tubes) - Microbial DNA Free (50/pk)